Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Vials with screw thread, Nalgene®

Vials with screw thread, Nalgene®

Serum vial, 20,00 ml, Clear, No, 20-415, +
  215-1499
   Low Manufacturing Impact
Vials with screw thread, Nalgene®
  215-1499
   Low Manufacturing Impact
Vials with screw thread, Nalgene®
Vials with screw thread, Nalgene®

Serum vial, 20,00 ml, Clear, No, 20-415, +

 
17 820,00 Kč
   
1 * 612 KS
 

These vials are ideal for diagnostic and biological sera, pharmaceuticals for veterinary and ophthalmologic usage as well as many other applications. They are made from clear or coloured brown PETG.

  • Available clear or coloured brown, sterile or non-sterile
  • A wide range of different screw caps is optionally available
  • Manufactured and packed in workrooms equipped with HEPA filters

Closures for vials: HDPE, coloured. For safe and tight closure of narrow-neck vials. Available in non-sterile or sterile versions. Non sterile caps are loose packed, sterile caps are loose packed in a box with a three-section insert.

For safe and tight closure of narrow-neck vials.

Documents

 
  • Cap included:
    No
  • Cap Size:
    20-415
  • Sterile:
    +
  • Capacity:
    20,00 ml
  • Colour:
    Clear
  • Description:
    Serum vial
  • Cat. No.:
    215-1499
  • Environmental certification:
    NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.