Malonic acid ≥99%
Glass bottle for solids, 100 g
Dodavatel: Thermo Fisher Scientific
Přejít na:
Product Details & Documents
Purity: 99% CAS No.: 141-82-2 Molecular Formula: C3H4O4 Formula Weight: 104.06 Flash Point: 157°(314°F) Melting Point: 132-136° dec. Beilstein Reference: 1751370 Merck Reference: 14,5710 MDL No.: MFCD00002707
Synonyma: 1,3-Propanedioic acid
- Číslo CAS:141-82-2
- Vzorec:C₃H₄O₄
- MDL číslo:MFCD00002707
- Molecular Weight:104.06 g/mol
- Merck Index:14,5710
- Bod varu:387 °C (1013 hPa)
- Bod tání:132…136 °C (dec.)
- Hustota:1.62
- Bod vzplanutí:157 °C (314 °F)
- Teplota pro skladování:Ambient
- EINECS:205-503-0
Documents
Specifikace
Specifikace
- Pack type:Glass bottle for solids
- Pk:100 g
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



