cis-2,6-Dimethylmorpholine ≥97%
Glass bottle for solids, 10 g
Dodavatel: Thermo Fisher Scientific
cis-2,6-Dimethylmorpholine ≥97%
Katalogové číslo L13007.09
Dodavatel: Thermo Fisher Scientific
Číslo CAS: 6485-55-8
Přejít na:
Product Details & Documents
Purity: 97% CAS No.: 6485-55-8 Molecular Formula: C6H13NO Formula Weight: 115.18 Flash Point: 48°(118°F) Refractive Index: 1.4460 Melting Point: -85° Boiling Point: 146-147° Beilstein Reference: 878182 UN No.: 1992 MDL No.: MFCD00078428
- Číslo CAS:6485-55-8
- Vzorec:C₆H₁₃NO
- MDL číslo:MFCD00078428
- Molecular Weight:115.18 g/mol
- Bod varu:146…147 °C
- Bod tání:–85 °C
- Hustota:0.935
- Bod vzplanutí:48 °C (118 °F)
- Teplota pro skladování:Ambient
- EINECS:229-353-0
- UN:1992
- ADR:3,III
Documents
Specifikace
Specifikace
- Pack type:Glass bottle for solids
- Pk:10 g
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



