MICONAZOLE NITRATE IMP. C (EP) AS NITRAT 1 * 100 mg
Catalog # PROOMM0281.03
Supplier: LGC Standards PROMOCHEM
MICONAZOLE NITRATE IMP. C (EP) AS NITRAT 1 * 100 mg
Catalog # PROOMM0281.03
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
2-Propanol System Suitability, Reference Standard
Supplier: USP