Benzo(a)anthracene D12 10 ng/µl 1 * 10 mL
Catalog # EHERL20545100CY
Supplier: EHRENSTORFER
Frequently Bought Together
Conductivity standards, KCl, referenced at 25 °C
Supplier: REAGECON
Accessories for perfectION™ electrodes
Supplier: Mettler - Toledo
Neomycin sulphate
Supplier: EHRENSTORFER
CEFAZOLIN SODIUM SALT 1 * 75 mg
Supplier: Merck
O-Acetylsalicylic acid
Supplier: EHRENSTORFER
Capric Alcohol, Reference Standard
Supplier: USP
Deltamethrin certified 1 * 250 mg
Supplier: EHRENSTORFER
Sulfamethoxazole, Reference Standard
Supplier: USP
Buffer Solution pH 6.00 (20°C) 1 * 1.000 mL
Supplier: PanReac AppliChem
1-Bromo-2-chloroethane
Supplier: EHRENSTORFER
Trace metals - paint chips, (CRM), Supelco®
Supplier: Merck
Oxyquinoline Sulfate, Reference Standard
Supplier: USP
Total organic carbon (TOC) standards
Supplier: REAGECON
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
SULFAMETHOXAZOLE-D4 1 * 5 mg
Supplier: LGC Standards PROMOCHEM
Conductivity standards, KCl, referenced at 25 °C
Supplier: Thermo Fisher Scientific