Isazofos 10 ng/µl 1 * 10 mL
Catalog # EHERL14377000IO
Supplier: EHRENSTORFER
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
VWR® ARISTAR®, Multi-Element Calibration Standard IV in dil. Nitric Acid for ICP
Supplier: VWR Chemicals
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Sucrose (Brix) standards, stabilised solutions for refractometry
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Benzoic acid, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
pH buffer, single-use, SINGLET™
Supplier: Hach