ContactUsLinkComponent
Micro-Resistance thermometer W 5780 NN HT. Glass shaft, sensor Pt 1000, 2 x 4 mm banana plugs, length 250 mm, 6 mm Ø, -30...+200 °C 1 * 1 items
Catalog # 442-0543
Supplier:  SI Analytics
Micro-Resistance thermometer W 5780 NN HT. Glass shaft, sensor Pt 1000, 2 x 4 mm banana plugs, length 250 mm, 6 mm Ø, -30...+200 °C 1 * 1 items
Catalog # 442-0543
Supplier:  SI Analytics
Frequently Bought Together