Orders LinkContactUsLinkComponent
Empty solid phase extraction columns, CHROMABOND®
Empty solid phase extraction columns, CHROMABOND®
Empty LV PP columns with PE frits, for 200/500 mg adsorbent weight, 15 ml
Catalog # MANA732501
Supplier:   MACHEREY-NAGEL
Empty solid phase extraction columns, CHROMABOND®
Catalog # MANA732501
Supplier:  MACHEREY-NAGEL
Empty solid phase extraction columns, CHROMABOND®
Empty solid phase extraction columns, CHROMABOND®
Empty LV PP columns with PE frits, for 200/500 mg adsorbent weight, 15 ml
Price
5 860,00 Kč
Log in for availability
1 * 50 KS
Product Details & Documents
Empty and reservoir columns with accessories for solid phase extraction.
  • Empty PP and glass columns for individual packing with CHROMABOND® or other adsorbents
  • Reservoir columns from PP for application of medium-size samples
  • Accessories for SPE columns (filter elements, adapters)
Specifications
  • Capacity:
    15 ml
  • Description:
    Empty LV PP columns with PE frits, for 200/500 mg adsorbent weight
  • Seq:
    12
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • PE frits for LV PP columns 15 ml, for 100 mg adsorbent weight
    PE frits for LV PP columns 15 ml, for 100 mg adsorbent weight
    MANA732019
    Log in for availability
    1 * 250 KS - 3 830,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weight
    PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weight
    MANA732020
    Log in for availability
    1 * 250 KS - 5 370,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • Adapters (PP) for PP columns (1, 3 and 6 ml)
    Adapters (PP) for PP columns (1, 3 and 6 ml)
    MANA730101
    Log in for availability
    1 * 10 KS - 2 790,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • Adapters (PVDF) for glass columns (3 and 6 ml)
    Adapters (PVDF) for glass columns (3 and 6 ml)
    MANA730105
    Log in for availability
    1 * 10 KS - 5 800,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 70 ml
    PE frits for PP columns, 70 ml
    MANA730026
    Log in for availability
    1 * 250 KS - 8 150,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • Glass fiber frits for glass columns, 6 ml
    Glass fiber frits for glass columns, 6 ml
    MANA730192
    Log in for availability
    1 * 250 KS - 6 910,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 15 ml
    PE frits for PP columns, 15 ml
    MANA730351
    Log in for availability
    1 * 250 KS - 6 910,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 45 ml
    PE frits for PP columns, 45 ml
    MANA730356
    Log in for availability
    1 * 250 KS - 7 920,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 150 ml
    PE frits for PP columns, 150 ml
    MANA730475
    Log in for availability
    1 * 250 KS - 8 760,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 30 ml
    PE frits for PP columns, 30 ml
    MANA730034
    Log in for availability
    1 * 250 KS - 7 490,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • Adapters (PE) for PP columns (30 and 70 ml)
    Adapters (PE) for PP columns (30 and 70 ml)
    MANA730566
    Log in for availability
    1 * 1 KIT - 844,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • Adapters (PE) for PP columns (15, 45 and 70 ml)
    Adapters (PE) for PP columns (15, 45 and 70 ml)
    MANA730385
    Log in for availability
    1 * 10 KS - 3 630,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • Glass fiber frits for glass columns, 3 ml
    Glass fiber frits for glass columns, 3 ml
    MANA730191
    Log in for availability
    1 * 250 KS - 5 690,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 1 ml
    PE frits for PP columns, 1 ml
    MANA730164
    Log in for availability
    1 * 250 KS - 3 770,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 6 ml
    PE frits for PP columns, 6 ml
    MANA730163
    Log in for availability
    1 * 250 KS - 6 560,00 Kč
Product Information
  • Catalog #
    Availability
    Price
  • PE frits for PP columns, 3 ml
    PE frits for PP columns, 3 ml
    MANA730162
    Log in for availability
    1 * 250 KS - 5 340,00 Kč