ContactUsLinkComponent
EC HPLC column EC 250/4 NUCLEOSIL 100-7 C8 Length: 250 mm, ID: 4 mm pack of 1 1 * 1 items
Catalog # MANA720017.40
Supplier:  MACHEREY-NAGEL
EC HPLC column EC 250/4 NUCLEOSIL 100-7 C8 Length: 250 mm, ID: 4 mm pack of 1 1 * 1 items
Catalog # MANA720017.40
Supplier:  MACHEREY-NAGEL
Frequently Bought Together