ContactUsLinkComponent
Extraction thimbles MN 645 W Format:31x205mm (Inner diameterxHeight) pack of 25. 1 * 25 items
Catalog # MANA645126
Supplier:  MACHEREY-NAGEL
Extraction thimbles MN 645 W Format:31x205mm (Inner diameterxHeight) pack of 25. 1 * 25 items
Catalog # MANA645126
Supplier:  MACHEREY-NAGEL
Frequently Bought Together