ContactUsLinkComponent
Filter paper folded MN 675 1/4 45cm diameter, pack of 100. 1 * 100 items
Catalog # MANA548045
Supplier:  MACHEREY-NAGEL
Filter paper folded MN 675 1/4 45cm diameter, pack of 100. 1 * 100 items
Catalog # MANA548045
Supplier:  MACHEREY-NAGEL
Frequently Bought Together