Filter paper folded MN 660 1/4 27cm diameter, pack of 100. 1 * 100 items
Catalog # MANA547027
Supplier: MACHEREY-NAGEL
Filter paper folded MN 660 1/4 27cm diameter, pack of 100. 1 * 100 items
Catalog # MANA547027
Supplier: MACHEREY-NAGEL
Frequently Bought Together
VWR®, Syringe Filters
Supplier: VWR Collection
Bottle top vacuum filtration systems, VacuCap®, Supor® membrane
Supplier: Cytiva (Formerly Pall Lab)
Glass microfibre filters without binder, grade GF/C, Whatman™
Supplier: Whatman products (Cytiva)
VWR® Weighing Paper, Grade 531
Supplier: VWR Collection
Membrane filters, cellulose nitrate, non sterile, Whatman™
Supplier: Whatman products (Cytiva)
Syringe filters, Acrodisc®, Supor® membrane
Supplier: Cytiva (Formerly Pall Lab)
Membrane filters, type 113
Supplier: Sartorius
Accessories for MA series
Supplier: Sartorius Balances
VWR®, Qualitative Filter Papers
Supplier: VWR Collection
Vacuum filtration units, Rapid-Flow™ filters MF 75, Nalgene®
Supplier: Thermo Fisher Scientific
MicroFunnel™ Filter Funnels
Supplier: Cytiva (Formerly Pall Lab)