ContactUsLinkComponent
Filter paper folded MN 619 1/4 27cm diameter, pack of 100. 1 * 100 items
Catalog # MANA537027
Supplier:  MACHEREY-NAGEL
Filter paper folded MN 619 1/4 27cm diameter, pack of 100. 1 * 100 items
Catalog # MANA537027
Supplier:  MACHEREY-NAGEL
Frequently Bought Together