Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Buffer, TE solution (TRIS-EDTA buffer) pH 8,0

Buffer, TE solution (TRIS-EDTA buffer) pH 8,0

TE buffer solution (TRIS-EDTA buffer) pH 8,0, Glass bottle, 1 L
Catalog # A0386.1000
Supplier:   PanReac AppliChem
Buffer, TE solution (TRIS-EDTA buffer) pH 8,0
Catalog # A0386.1000
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, TE solution (TRIS-EDTA buffer) pH 8,0

TE buffer solution (TRIS-EDTA buffer) pH 8,0, Glass bottle, 1 L

CAS Number:  
Price
1 320,00 Kč
Log in for availability
1 * 1 L
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TE buffer solution (TRIS-EDTA buffer) pH 8,0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.