Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Lysis buffer, CTAB

Lysis buffer, CTAB

CTAB, lysis buffer, 500 mL
Catalog # A4150.0500
Supplier:   PanReac AppliChem
Lysis buffer, CTAB
Catalog # A4150.0500
Supplier:  PanReac AppliChem
CAS Number:  
Lysis buffer, CTAB
Lysis buffer, CTAB

CTAB, lysis buffer, 500 mL

CAS Number:  
Price
1 160,00 Kč
Log in for availability
1 * 500 mL
Product Details & Documents

Cationic detergent cetyltrimethylammonium bromide (CTAB) is used to liberate and complex with total cellular nucleic acids.

  • Form: Liquid
  • Storage temperature: Ambient
  • Specification pH (20 °C): 8,0 ±0,1

This buffer contains 2% w/v CTAB, 20 mM EDTA.Na₂.2H₂O, 1,4 M sodium chloride and 100 mM tris ultrapure. Effective for removing polysaccharides, phenolic compounds and other enzyme-inhibiting contaminants found in plant cells in the supernatant.

Documents

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    CTAB, lysis buffer

Specifications