ContactUsLinkComponent
Areometer, BS718, M50 series, measuring range:1.050-1.100:0.001g/ml, without thermometer, length:270mm, reference temperature:20°C, suitable for government verification. 1 * 1 items
Catalog # AMARH808018
Supplier:  Amarell
Areometer, BS718, M50 series, measuring range:1.050-1.100:0.001g/ml, without thermometer, length:270mm, reference temperature:20°C, suitable for government verification. 1 * 1 items
Catalog # AMARH808018
Supplier:  Amarell
Frequently Bought Together