Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
BUFFER PH 7.00 20°C YELLOW 1 * 5 L

BUFFER PH 7.00 20°C YELLOW 1 * 5 L

Catalog # REGEBPH03
Supplier:   REAGECON
BUFFER PH 7.00 20°C YELLOW 1 * 5 L
Catalog # REGEBPH03
Supplier:  REAGECON
BUFFER PH 7.00 20°C YELLOW 1 * 5 L
Price
4 260,00 Kč
Log in for availability
1 * 5 L
Product Details & Documents