96 Well Filter Plate, ACROPREP 96 0.2UM PTFE Membrane, 350ul, Natural 1 * 10 items
516-6887
96 Well Filter Plate, ACROPREP 96 0.2UM PTFE Membrane, 350ul, Natural 1 * 10 items
516-6887
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection