ContactUsLinkComponent
pH combination electrode with screw plug head S 9090. Metal combination electrode Pt 8281 HD. Low maintenance redox-combination elec trode, glass shaft, KPG annular gap diaphragm, Referid®-electrolyte, Silamid®-reference system, sensor platinum disk 1 * 1 items
Catalog # SCOI285112414
Supplier:  SI Analytics
pH combination electrode with screw plug head S 9090. Metal combination electrode Pt 8281 HD. Low maintenance redox-combination elec trode, glass shaft, KPG annular gap diaphragm, Referid®-electrolyte, Silamid®-reference system, sensor platinum disk 1 * 1 items
Catalog # SCOI285112414
Supplier:  SI Analytics
Frequently Bought Together