ContactUsLinkComponent
SONOREX LONGLIFE RK 515 CH, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 280X234X300 MM 19,7 L, OSCILLATING TANK OF 2 MM S/S AISI 316 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C (86-176F), DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 1200 1 * 1 items
Catalog # BAND3035
SONOREX LONGLIFE RK 515 CH, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 280X234X300 MM 19,7 L, OSCILLATING TANK OF 2 MM S/S AISI 316 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C (86-176F), DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 1200 1 * 1 items
Catalog # BAND3035
Frequently Bought Together