SONOREX LONGLIFE RK 103 CH, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 220X135X150 MM 4,5 L, OSCILLATING TANK OF 2 MM S/S AISI 3 16 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C (86-176F), DRAIN BALL COCK 1/4 , ULTRASONIC PEAK OUTPUT 560 W 1 * 1 items
Catalog # BAND3032
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together