BUFFER SOLUTION PH 7,00 1 * 500 mL
Catalog # METR6.2307.110
Supplier: METROHM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
UV/Visible standards, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
COD calibration standard solutions (NIST traceable)
Supplier: Hach
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond