4,4´-DDE 10 ng/µl 1 * 10 mL
Catalog # EHERL12041000AL
Supplier: EHRENSTORFER
Frequently Bought Together
MCPB D6 (ring D3, methyl D3) 100 ng/µl 1 * 1,1 mL
Supplier: EHRENSTORFER
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
o-Anisidine
Supplier: EHRENSTORFER
VWR® ARISTAR®, Holmium Standard Solutions for ICP-MS
Supplier: VWR Chemicals
BAICALIN, PHYPROOF(R) REFERENCE SUBSTANCE 1 * 10 mg
Supplier: Merck
GINSENOSIDE RB2 1 * 10 mg
Supplier: Merck
Triethyl Citrate, Reference Standard
Supplier: USP
Pesticide reference standard, ε-BHC 100 µg/ml in methanol, Dr Ehrenstorfer
Supplier: EHRENSTORFER
Acetate Buffer pH 2.5 CI270 1 * 10 L
Supplier: REAGECON
Buffer Solution pH 9.180 1 * 1 L
Supplier: PanReac AppliChem
Ethylene glycol, Supelco®
Supplier: Merck
TOC standard, according to ISO/CEN EN1484, Supelco®
Supplier: Merck