SCREW-JOINT 1 * 1 items
Catalog # IKAA1582800
Supplier: IKA
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Oxalic acid 0.5 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Accessories for PIPETBOY acu / pro
Supplier: Integra
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG