ContactUsLinkComponent
20mm silicone/ptfe seal 3mm thick,ultraclean, for working with high temperatures upto 250°c. **available in packs of 100** 1 * 100 items
Catalog # 548-0271
Supplier:  CHROMACOL
20mm silicone/ptfe seal 3mm thick,ultraclean, for working with high temperatures upto 250°c. **available in packs of 100** 1 * 100 items
Catalog # 548-0271
Supplier:  CHROMACOL
Frequently Bought Together