Caps for bottles, open top, Microlink®, WHEATON®
Screw cap, solid top, with PP/silicone/PTFE liner, 38-430, White
Katalogové číslo WHEA240235-01
Dodavatel: DWK Life Sciences
Caps for bottles, open top, Microlink®, WHEATON®
Katalogové číslo WHEA240235-01
Dodavatel: DWK Life Sciences
Přejít na:
Product Details & Documents
Screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
- PTFE or PP on the exposed face of the liner imparts an inert, chemical-resistant surface
- Open top caps provide access with a syringe
Ultrasonically welded liner to the cap eliminates the possibility of glue contamination and bond failure on autoclaving. Low-compression-set silicone provides prolonged period of time sample storage over wide temperature ranges.
Specifikace
- Thread:38-430
- Colour:White
- Description:Screw cap, solid top, with PP/silicone/PTFE liner
- Seq:14