Orders LinkContactUsLinkComponent
Well visualisation plates
Well visualisation plates
Well visualisation plate, 9.5×11.6 cm, with 4 rails for locking portrait and landscape gels in place
Katalogové číslo 730-2202
Dodavatel:   EMBITEC
Well visualisation plates
Katalogové číslo 730-2202
Dodavatel:  EMBITEC
Well visualisation plates
Well visualisation plates
Well visualisation plate, 9.5×11.6 cm, with 4 rails for locking portrait and landscape gels in place
Price
363,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
RunOne™ well visualisation plates are designed to make gel loading simple. The light colour of most gel units provides little or no contrast for the agarose gel and the gel wells. RunOne™ well visualisation plates are red, fit securely into the RunOne™ system, and provide a stable platform for gel running.
Specifications
  • Description:
    Well visualisation plate, 9.5×11.6 cm, with 4 rails for locking portrait and landscape gels in place
  • Seq:
    02