Orders LinkContactUsLinkComponent
Gel casting system, MultiCaster
Gel casting system, MultiCaster
UVT tray for MultiCaster
Catalog # 730-0360
Supplier:   EMBITEC
Gel casting system, MultiCaster
Catalog # 730-0360
Supplier:  EMBITEC
Gel casting system, MultiCaster
Gel casting system, MultiCaster
UVT tray for MultiCaster
Price
1 750,00 Kč
Log in for availability
1 * 2 KS
Product Details & Documents
Moulded PC. Compatible with the RunOne™ electrophoresis system.
  • Simple to use - no gaskets, clamps or tape required
  • Flexible with options for well size, well configuration, number of wells and resolving area
Specifications
  • Description:
    UVT tray for MultiCaster
  • Seq:
    05
  • Cat. No.:
    730-0360
  • Gel dimensions:
Recommendations