GRAM-HÜCKER L slow action kit The L (slow action) kit is advised for individual staining of slides. Kit Components Cristal Violet Oxalate: 1 x 240 ml Lugol, PVP-stabilized Solution: 1 x 240 ml Fast Differentiator (Alcohol/Acetone) or Slow Different 1 * 1 KIT
Catalog # 720-1188
Supplier: RAL
Frequently Bought Together