Test Strips, Iron, 0 - 5 mg/l, 25 tests, Steps: 0, 0.15, 0.3, 0.6, 1, 2, 5 mg/l, 1 * 25 items
Catalog # HACH2745325
Supplier: Hach
Test Strips, Iron, 0 - 5 mg/l, 25 tests, Steps: 0, 0.15, 0.3, 0.6, 1, 2, 5 mg/l, 1 * 25 items
Catalog # HACH2745325
Supplier: Hach
Frequently Bought Together
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF