Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Chloroform ≥98% stabilised, TECHNICAL

Chloroform ≥98% stabilised, TECHNICAL

Metal drum, 25 L
Catalog # 22720.462
Health
Toxic
Supplier:   VWR Chemicals
Chloroform ≥98% stabilised, TECHNICAL
Catalog # 22720.462
Supplier:  VWR Chemicals
CAS Number:  67-66-3
Chloroform ≥98% stabilised, TECHNICAL
Chloroform ≥98% stabilised, TECHNICAL

Metal drum, 25 L

CAS Number:  67-66-3
Price
Inquire For Price
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
  • CAS Number:67-66-3
  • Formula:CHCl₃
  • MDL Number:MFCD00000826
  • Molecular Weight:119.38 g/mol
  • Merck Index:13,02160
  • Boiling Pt:61.7 °C (1013 hPa)
  • Melting Pt:–63 °C
  • Density:1.4832 g/cm³ (20 °C)
  • EINECS:200-663-8
  • ADR:6.1,III
  •  :Stabilised with ethanol 0.6 %
  • Storage Temperature:Ambient
  • UN:1888
  • REACH:01-2119486657-20
Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Density (20/4):
    1.473 - 1.489
  • Ethanol:
    0.1 - 1.0 %

Specifications

  • Pack type:
    Metal drum
  • Pk:
    25 L

Specifications