Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

D-(-)-Sorbitol, TECHNICAL

Plastic bucket, 5 kg
Catalog # 28210.367
Supplier:   VWR Chemicals
D-(-)-Sorbitol, TECHNICAL
Catalog # 28210.367
Supplier:  VWR Chemicals
CAS Number:  50-70-4
D-(-)-Sorbitol, TECHNICAL
D-(-)-Sorbitol, TECHNICAL

Plastic bucket, 5 kg

CAS Number:  50-70-4
Price
6 200,00 Kč
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,08797
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:94…98 °C
  • Density:1.524 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:200-061-5
Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.