Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Ethyl acetate ≥99%, TECHNICAL

Ethyl acetate ≥99%, TECHNICAL

Metal drum, 200 L
Catalog # 23879.557
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   VWR Chemicals
Ethyl acetate ≥99%, TECHNICAL
Catalog # 23879.557
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  141-78-6
Ethyl acetate ≥99%, TECHNICAL
Ethyl acetate ≥99%, TECHNICAL

Metal drum, 200 L

CAS Number:  141-78-6
Price
Inquire For Price
Log in for availability
1 * 200 L
Product Details & Documents
Synonyms:  Acetic acid ethyl ester
  • CAS Number:141-78-6
  • Formula:CH₃COOC₂H₅
  • MDL Number:MFCD00009171
  • Molecular Weight:88.11 g/mol
  • Merck Index:13,03792
  • Boiling Pt:77.1 °C (1013 hPa)
  • Melting Pt:–83 °C
  • Density:0.902 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Storage Temperature:Ambient
  • EINECS:205-500-4
  • UN:1173
  • ADR:3,II
  • REACH:01-2119475103-46
Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Appearance:
    Clear colourless liquid
  • IR Spectrum:
    Passes test

Specifications

  • Pack type:
    Metal drum
  • Pk:
    200 L
  • Cat. No.:
    23879.557
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.