PISTON WITH COUPLING PTFE 20 ML 1 * 1 items
Catalog # METR6.1556.220
Supplier: METROHM
Frequently Bought Together
Hydrochloric acid 20%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Combined pH electrodes, Metrosensor
Supplier: METROHM
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Sedimentation cones, Imhoff type
Supplier: VITLAB
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
Beakers, tall form, DURAN®
Supplier: DWK Life Sciences
ABSAUGSPITZE M6 ID/AD 2/2,9MM, L 151MM 1 * 1 ST
Supplier: METROHM
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
CYLINDER 20 ML CLEAR GLASS 1 * 1 items
Supplier: METROHM
VWR® High Performance, Bottles, Wide Neck with Screw Cap
Supplier: VWR Collection
PISTON WITH COUPLING PTFE 50ML 1 * 1 items
Supplier: METROHM
TUBING PTFE 1 * 1 items
Supplier: METROHM