Orders LinkContactUsLinkComponent
Gel casting system, MultiCaster
Gel casting system, MultiCaster
MultiCaster, yellow casting stand, UVT tray, 8 reversible combs, and a red plate, 12-channel pipette compatible
Catalog # 700-4210
Supplier:   EMBITEC
Gel casting system, MultiCaster
Catalog # 700-4210
Supplier:  EMBITEC
Gel casting system, MultiCaster
Gel casting system, MultiCaster
MultiCaster, yellow casting stand, UVT tray, 8 reversible combs, and a red plate, 12-channel pipette compatible
Price
10 510,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Moulded PC. Compatible with the RunOne™ electrophoresis system.
  • Simple to use - no gaskets, clamps or tape required
  • Flexible with options for well size, well configuration, number of wells and resolving area
Specifications
  • Description:
    MultiCaster, yellow casting stand, UVT tray, 8 reversible combs, and a red plate, 12-channel pipette compatible
  • Seq:
    04
  • Cat. No.:
    700-4210
  • Gel dimensions:
    115×89