MICROSYRINGE 10µL GAS ANALYSIS MS/GF FIXED NEEDLE 1 * 1 items
Catalog # 613-1742
Supplier: Hospidex
Frequently Bought Together
QuEChERS mixes, clean-up, CHROMABOND®
Supplier: MACHEREY-NAGEL
PK36 100ML CLEAR SERUM BOTTLE 51.7X94.5MM 1 * 36 items
Supplier: Merck
Microliter™ 700 series syringes
Supplier: HAMILTON BONADUZ
Capillary columns, CP-Sil
Supplier: VARIAN
METAGUARD 2.0MM PURSUIT XRS 3U C18,3 1 * 1 items
Supplier: VARIAN
HPLC columns, Cogent UDC-Cholesterol™
Supplier: MICROSOLV
HPLC waste-sets, V2.0
Supplier: S.C.A.T.
COLUMN CHIRALPAK IM 250X4.6MM 5M 1 * 1 items
Supplier: CHIRAL TECHNOLOGIES
SkillPak DEAE-650M 5x1mL col., 7.0x25mm 1 * 1 items
Supplier: TOSOH Bioscience
Spare needles for autosampler syringes
Supplier: Agilent