ContactUsLinkComponent
Sensorturn Rotary speed range 14 - 110 rpm includes IR-Sensor and connector for foot pedal. 5 operating modes: .IR-Sensor: Time / Constant .Foot pedal: Standard / Start-Stop / Time .Turntable with transparent silicone covering & centring ring 1 * 1 items
Catalog # 451-1386
Supplier:  WARTEWIG
undefined
Sensorturn Rotary speed range 14 - 110 rpm includes IR-Sensor and connector for foot pedal. 5 operating modes: .IR-Sensor: Time / Constant .Foot pedal: Standard / Start-Stop / Time .Turntable with transparent silicone covering & centring ring 1 * 1 items
Catalog # 451-1386
Supplier:  WARTEWIG
Frequently Bought Together