Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Wipes for general applications, Polytex®

75 g/m², Blue, 420×420 mm, Pack of 35
Catalog # 115-2703
Wipes for general applications, Polytex®
Catalog # 115-2703
Wipes for general applications, Polytex®
Wipes for general applications, Polytex®

75 g/m², Blue, 420×420 mm, Pack of 35

Price
153,00 Kč
Log in for availability
1 * 35 KS
Product Details & Documents

Soft cloths for wet wiping of sensitive surfaces.

  • 100% PP, reusable
  • Solvent resistant
  • Anti-static and fluff-free for universal lab applications and food areas
  • High tear resistance and good absorption capacity
Specifications
  • Packed:
    Pack of 35
  • Colour:
    Blue
  • Description:
    75 g/m²
  • L×W:
    420×420 mm