ContactUsLinkComponent
Metal combination electrode with screw plug head Pt 9080. Glass shaft, ceramic diaphragm, electrolyte KCl 3 mol/l, Ag/AgCl-reference system, sensor platinum ring 6 mm Ø, refilling tube, plug head, length 120 mm, 12 mm Ø, -5...+100 °C 1 * 1 items
Catalog # SCOI285111089
Supplier:  SI Analytics
Metal combination electrode with screw plug head Pt 9080. Glass shaft, ceramic diaphragm, electrolyte KCl 3 mol/l, Ag/AgCl-reference system, sensor platinum ring 6 mm Ø, refilling tube, plug head, length 120 mm, 12 mm Ø, -5...+100 °C 1 * 1 items
Catalog # SCOI285111089
Supplier:  SI Analytics
Frequently Bought Together