ContactUsLinkComponent
Metal combination electrode Pt 7781 HD. Low maintenance, glass shaft, ceramic diaphragm, gel electrolyte, Ag/AgCl-reference system, sensor platinum pin 1 mm Ø, screw plug head, length 120 mm, 12 mm Ø, -5...+60 °C 1 * 1 items
Catalog # SCOI285110754
Supplier:  SI Analytics
Metal combination electrode Pt 7781 HD. Low maintenance, glass shaft, ceramic diaphragm, gel electrolyte, Ag/AgCl-reference system, sensor platinum pin 1 mm Ø, screw plug head, length 120 mm, 12 mm Ø, -5...+60 °C 1 * 1 items
Catalog # SCOI285110754
Supplier:  SI Analytics
Frequently Bought Together