Diethylether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
5190, 5 L
Supplier: Avantor
Diethylether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 8254.5000
Supplier: Avantor
CAS Number: 60-29-7
Jump to:
Product Details & Documents
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12
- Merck Index:12,03852
- Boiling Pt:35 °C (1013 hPa)
- Melting Pt:–123 °C
- Density:0,7135 g/cm³ (20 °C)
- Flash Pt:–40 °C
- UN:1155
- ADR:3,I
- :Density: 0,7135 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-467-2
- REACH:01-2119535785-29
Documents
Specifications
Specification Test Results
- Assay (by GC):min. 99,5 %
- Alcohol (C2H5OH):max. 0,01 %
- Carbonyl Compounds (as HCHO):max. 0,001 %
- Color (APHA):max. 10
- Density (g/ml) at 20°C:0.712-0.714
- Foreign Odor:passes test
- Peroxide (as H202):max. 3 ppm
- Residue after Evaporation:max. 0,001 %
- Substances Darkened by H2SO4:passes test
- Titrable Acid (meq/g):max. 0,0002
- Water (H2O):max. 0,01 %
- Trace Impurities (in ppm):-
- Aluminium (Al):max. 0,5
- Barium (Ba):max. 0,1
- Boron (B):max. 0,02
- Cadmium (Cd):max. 0,05
- Calcium (Ca):max. 0,5
- Chromium (Cr):max. 0,02
- Cobalt (Co):max. 0,02
- Copper (Cu):max. 0,02
- Iron (Fe):max. 0,1
- Lead (Pb):max. 0,1
- Magnesium (Mg):max. 0,1
- Manganese (Mn):max. 0,02
- Nickel (Ni):max. 0,02
- Tin (Sn):max. 0,1
- Zinc (Zn):max. 0,1
Specifications
- Bal.:5 L
- Sekvence:5190
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



