Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Směs pro test HPLC kolony s reverzní fází v acetonitrilu, 1ml 1 * 1 mL
Catalog # MANA722394
Dodavatel:
MACHEREY-NAGEL
Expand image
Směs pro test HPLC kolony s reverzní fází v acetonitrilu, 1ml 1 * 1 mL
Catalog # MANA722394
Dodavatel:
MACHEREY-NAGEL
Frequently Bought Together
Diluid 100 Plus for hematology (max. 0.001% acetone), J.T.Baker®
Dodavatel: Avantor
Reagents for DR 2800, DR 3800 DR 3900, DR 5000 and DR 6000
Dodavatel: Hach
Stain,3 Step-Solution B,1GAL,3800ML 1 * 1 KS
Dodavatel: SHANDON