Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Stand bases

Stand bases

2,2 kg, 150×300×6 mm
Catalog # 241-3167
Supplier:   BOCHEM
Stand bases
Catalog # 241-3167
Supplier:  BOCHEM
Stand bases
Stand bases

2,2 kg, 150×300×6 mm

Price
1 080,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents

Steel, varnished.

  • With M10 threaded hole
  • One adjustable foot

Certifications: DIN 12892

Specifications
  • W×D×H:
    150×300×6 mm
  • Weight:
    2,2 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.