ContactUsLinkComponent
Areometer, casagrande, DIN18123, measuring range:0.995-1.030:0.0005g/cm , length:350mm, reference temperature:20°C, suitable for gov ernment verification. 1 * 1 items
Catalog # 614-0532
Supplier:  Amarell
Areometer, casagrande, DIN18123, measuring range:0.995-1.030:0.0005g/cm , length:350mm, reference temperature:20°C, suitable for gov ernment verification. 1 * 1 items
Catalog # 614-0532
Supplier:  Amarell
Frequently Bought Together