ContactUsLinkComponent
Pipette 135mm lg 1.5ml,pipettes - soft polyethylene (ldpe) used as intake and pipette. due to the very soft and durable elastic head allows the liquid to absorb and accurately dispense drops. when filled to not pipette empties itself of the pipettes 1 * 100 items
Catalog # 612-0622
Supplier:  SEMADENI
Pipette 135mm lg 1.5ml,pipettes - soft polyethylene (ldpe) used as intake and pipette. due to the very soft and durable elastic head allows the liquid to absorb and accurately dispense drops. when filled to not pipette empties itself of the pipettes 1 * 100 items
Catalog # 612-0622
Supplier:  SEMADENI
Frequently Bought Together