Ammonium citrate dibasic Ammonium citrate dibasic Ammonium citrate dibasic Ammonium citrate dibasic 1 * 500 g
Katalogové číslo SIAL25102-500G-R
Supplier: Merck
Ammonium citrate dibasic Ammonium citrate dibasic Ammonium citrate dibasic Ammonium citrate dibasic 1 * 500 g
Katalogové číslo SIAL25102-500G-R
Supplier: Merck
Frequently Bought Together
Citric acid, anhydrous ≥99.8%, powder, AnalaR NORMAPUR®
Supplier: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Ammonium formate ≥97%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium iodide 99.0-100.5% Ph. Eur.
Supplier: VWR Chemicals