S/Steel Ferrule Packed 10,Jour 1 * 10 items
Catalog # 554-1078
Supplier: VICI JOUR RESEARCH
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
VWR® Facial Tissues
Supplier: VWR Collection
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
VWR® SP100 Automatic Straw Pipette, Fixed Volume
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Titration beakers
Supplier: Mettler - Toledo
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
Accessories for titrators
Supplier: Mettler - Toledo
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
pH electrodes, SenTix® PLUS
Supplier: WTW
Electrode stand with swivel arm, Eutech
Supplier: EUTECH
VWR®, Hand Tally Counter
Supplier: VWR Collection
Plastic cuvette for LICO
Supplier: Hach
Ethanolamine ≥99.5% (by GC), AnalaR NORMAPUR® ACS analytical reagent
Supplier: VWR Chemicals