ContactUsLinkComponent
test sieve 200/50 mm, mesh = 20 mm ISO 3310-1 1 * 1 items
Catalog # 510-4145
Supplier:  FRITSCH
test sieve 200/50 mm, mesh = 20 mm ISO 3310-1 1 * 1 items
Catalog # 510-4145
Supplier:  FRITSCH
Frequently Bought Together