ContactUsLinkComponent
Cryoboxes with grids 9 x 9 places, PP yellow, format 133 x 133 x 52 mm, for cryo tubes 1,2 - 2 mL (Ø 12,8mm) 1 * 5 items
Catalog # 479-5166
Supplier:  RATIOLAB
Cryoboxes with grids 9 x 9 places, PP yellow, format 133 x 133 x 52 mm, for cryo tubes 1,2 - 2 mL (Ø 12,8mm) 1 * 5 items
Catalog # 479-5166
Supplier:  RATIOLAB
Frequently Bought Together