STAMMOPUR 23 - 1 LITRE, DISINFECTION AND CLEANING, TESTED ACCORDING TO THE GUIDELINES OF THE DGHM , CONCENTRATE, WITHOUT FORMALD EHYDE, CHLORINE, PHENOLS, BACTERICIDAL (INCL. TB.-B.), FUNGICIDAL, VIRUCIDAL (HBV/HIV, POLIO), NEUTRAL, PH 7, DOSAGE 1 * 1 l
Catalog # 142-2010
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together